missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Tantalum Standard for ICP 1000 ppm in 5% HNO3/HF, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010318250N
Description
- 1000 ppm in 5% HNO3/HF
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Natural Poly Bottle | |
| 91.97%,4.90%,0.98%,0.15% | |
| 1 | |
| F5Ta | |
| Approximately 0°C | |
| YRGLXIVYESZPLQ-UHFFFAOYSA-I | |
| pentafluorotantalum | |
| 250 mL | |
| High Purity |
| 7783-71-3 , 7697-37-2 , 7732-18-5 , 7783-71-3 | |
| 95.73%,5.10%,1.02%,0.15% | |
| Liquid | |
| UN2922 | |
| Standard | |
| F[Ta](F)(F)(F)F | |
| 275.94 | |
| 82218 |
Chemical Identifiers
| 7783-71-3 | |
| 275.94 | |
| 82218 | |
| F[Ta](F)(F)(F)F |
| F5Ta | |
| YRGLXIVYESZPLQ-UHFFFAOYSA-I | |
| pentafluorotantalum |