missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Tantalum (Ta) Standard for AAS 1000 ppm in 5% HNO3/1% HF, Ricca Chemical

Catalog No. RV010054100
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010054100 100 mL
RV010054500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010054100 Supplier Ricca Chemical Company Supplier No. RV010054100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 5% HNO3/1% HF
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7783-71-3 , 7697-37-2 , 7732-18-5 , 7783-71-3
Molecular Formula F5Ta
Molecular Weight (g/mol) 275.94
InChI Key YRGLXIVYESZPLQ-UHFFFAOYSA-I
PubChem CID 82218
IUPAC Name pentafluorotantalum
SMILES F[Ta](F)(F)(F)F

Specifications

Packaging Natural LDPE Bottle
CAS Min % 91.97%, 4.90%, 0.98%, 0.15%
CAS Max % 95.73%, 5.10%, 1.02%, 0.15%
Specific Gravity 1.01
Physical Form Liquid
UN Number UN2922
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity