missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Sodium (Na) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical

Catalog No. p-7074130
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010080100 100 mL
RV010080500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010080100 Supplier Ricca Chemical Company Supplier No. RV010080100N
Only null left

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7631-99-4 , 7697-37-2 , 7732-18-5
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
PubChem CID 24268
ChEBI CHEBI:63005
IUPAC Name sodium nitrate
SMILES [Na+].[O-][N+]([O-])=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 95.68%, 1.96%, 0.36%
CAS Max % 99.58%, 2.04%, 0.38%
Specific Gravity 1.011
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity