missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Ruthenium (Ru) Standard for AAS 1000 ppm in 2% HCl, Ricca Chemical

Catalog No. p-7074107
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010045100 100 mL
RV010045500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010045100 Supplier Ricca Chemical Company Supplier No. RV010045100C
Only null left

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 2% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 10049-08-8 , 7647-01-0 , 7732-18-5

Specifications

Packaging Amber Glass Bottle
CAS Min % 92.89%, 4.90%, 0.21%
CAS Max % 96.69%, 5.10%, 0.21%
Specific Gravity 1.045
Physical Form Liquid
UN Number UN3264
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity