missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Magnesium Standard for ICP/MS 100 ppm in 2% HNO3, Ricca Chemical

Catalog No. RV010533100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010533100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010533100 Supplier Ricca Chemical Company Supplier No. RV010533100N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 10377-60-3 , 7697-37-2 , 7732-18-5
Molecular Formula MgN2O6
Molecular Weight (g/mol) 148.31
MDL Number MFCD00011103
InChI Key YIXJRHPUWRPCBB-UHFFFAOYSA-N
PubChem CID 25212
ChEBI CHEBI:64736
IUPAC Name magnesium(2+) dinitrate
SMILES [Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 95.98%, 1.96%, 0.06%
CAS Max % 99.90%, 2.04%, 0.06%
Specific Gravity 1
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 100 mL
Grade High Purity
Chemical Name or Material Magnesium ICP-MS Standard
Show More Show Less