CAS RN 10377-60-3
Magnesium nitrate hydrate, Puratronic™, 99.999% (metals basis)
CAS: 10377-60-3 Molecular Formula: MgN2O6 Molecular Weight (g/mol): 148.31 MDL Number: MFCD00011103 InChI Key: YIXJRHPUWRPCBB-UHFFFAOYSA-N Synonym: magnesium nitrate,magnesium dinitrate,nitric acid, magnesium salt,unii-77cbg3un78,magnesium nitrate solution,nitric acid, magnesium salt 2:1,magniosan,hsdb 660,magnesium ii nitrate 1:2,magnesium standard for ic PubChem CID: 25212 ChEBI: CHEBI:64736 IUPAC Name: magnesium;dinitrate SMILES: [Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
VeriSpec™ Magnesium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
CAS: 10377-60-3 Molecular Formula: MgN2O6 Molecular Weight (g/mol): 148.31 MDL Number: MFCD00011103 InChI Key: YIXJRHPUWRPCBB-UHFFFAOYSA-N PubChem CID: 25212 ChEBI: CHEBI:64736 IUPAC Name: magnesium(2+) dinitrate SMILES: [Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
VeriSpec™ Magnesium Standard for ICP 10 g/L in 2% HNO3, Ricca Chemical
CAS: 10377-60-3 Molecular Formula: MgN2O6 Molecular Weight (g/mol): 148.31 MDL Number: MFCD00011103 InChI Key: YIXJRHPUWRPCBB-UHFFFAOYSA-N PubChem CID: 25212 ChEBI: CHEBI:64736 IUPAC Name: magnesium(2+) dinitrate SMILES: [Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
VeriSpec™ Magnesium (Mg) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
CAS: 10377-60-3 Molecular Formula: MgN2O6 Molecular Weight (g/mol): 148.31 MDL Number: MFCD00011103 InChI Key: YIXJRHPUWRPCBB-UHFFFAOYSA-N PubChem CID: 25212 ChEBI: CHEBI:64736 IUPAC Name: magnesium(2+) dinitrate SMILES: [Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
VeriSpec™ Magnesium Standard for ICP/MS 100 ppm in 2% HNO3, Ricca Chemical
CAS: 10377-60-3 Molecular Formula: MgN2O6 Molecular Weight (g/mol): 148.31 MDL Number: MFCD00011103 InChI Key: YIXJRHPUWRPCBB-UHFFFAOYSA-N PubChem CID: 25212 ChEBI: CHEBI:64736 IUPAC Name: magnesium(2+) dinitrate SMILES: [Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
VeriSpec™ Magnesium - Mg2+ Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical
CAS: 10377-60-3 Molecular Formula: MgN2O6 Molecular Weight (g/mol): 148.31 MDL Number: MFCD00011103 InChI Key: YIXJRHPUWRPCBB-UHFFFAOYSA-N PubChem CID: 25212 ChEBI: CHEBI:64736 IUPAC Name: magnesium(2+) dinitrate SMILES: [Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
VeriSpec™ Magnesium Nitrate Matrix Modifier for AAS 20 g/L in H2O, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
VeriSpec™ Magnesium Standard for ICP/MS 10 ppm in 2% HNO3, Ricca Chemical
CAS: 10377-60-3 Molecular Formula: MgN2O6 Molecular Weight (g/mol): 148.31 MDL Number: MFCD00011103 InChI Key: YIXJRHPUWRPCBB-UHFFFAOYSA-N PubChem CID: 25212 ChEBI: CHEBI:64736 IUPAC Name: magnesium(2+) dinitrate SMILES: [Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
VeriSpec™ Magnesium Nitrate Matrix Modifier for AAS 10,000 ppm in H2O, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility