missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Magnesium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical

Catalog No. RV010292100
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010292100 100 mL
RV010292250 250 mL
RV010292500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RV010292100 Supplier Ricca Chemical Company Supplier No. RV010292100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 10377-60-3 , 7697-37-2 , 7732-18-5
Molecular Formula MgN2O6
Molecular Weight (g/mol) 148.31
MDL Number MFCD00011103
InChI Key YIXJRHPUWRPCBB-UHFFFAOYSA-N
PubChem CID 25212
ChEBI CHEBI:64736
IUPAC Name magnesium(2+) dinitrate
SMILES [Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 95.44%, 1.96%, 0.60%
CAS Max % 99.34%, 2.04%, 0.62%
Specific Gravity 1
Physical Form Liquid
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Grade High Purity