missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Lanthanum (La) Standard for AAS 2% La in 2% HCl, Ricca Chemical

Catalog No. p-7073984
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010128100 100 mL
RV010128500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010128100 Supplier Ricca Chemical Company Supplier No. RV010128100N
Only null left

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 10099-58-8 , 7647-01-0 , 7732-18-5
Molecular Formula Cl3La
Molecular Weight (g/mol) 245.26
MDL Number MFCD00011068
InChI Key ICAKDTKJOYSXGC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[La+3]

Specifications

Packaging Natural LDPE Bottle
CAS Min % 92.58%, 3.46%, 1.96%
CAS Max % 96.36%, 3.60%, 2.04%
Specific Gravity 1.02
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity