missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Iridium Standard for ICP 1000 ppm in 10% HCl, Ricca Chemical

Catalog No. p-7073971
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010287250 250 mL
RV010287500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010287250 Supplier Ricca Chemical Company Supplier No. RV010287250N
Only null left

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 10% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 10025-83-9 , 7647-01-0 , 7732-18-5
Molecular Formula Cl3Ir
Molecular Weight (g/mol) 298.567
InChI Key DANYXEHCMQHDNX-UHFFFAOYSA-K
PubChem CID 25563
IUPAC Name trichloroiridium
SMILES Cl[Ir](Cl)Cl

Specifications

Packaging Natural Poly Bottle
CAS Min % 88.04%, 9.80%, 0.16%
CAS Max % 91.64%, 10.20%, 0.16%
Specific Gravity 1
Physical Form Liquid
UN Number UN1789
Melting Point Approximately 0°C
Type Standard
Quantity 250 mL
Grade High Purity