CAS RN 10025-83-9
CAS RN 10025-83-9
IUPAC Name:
iridium(3+) trichloride
Synonyms:
iridium(III) chloride
Molecular Weight (g/mol):
298.57
Molecular Formula:
Cl3Ir
InChi Key:
DANYXEHCMQHDNX-UHFFFAOYSA-K
SMILES:
[Cl-].[Cl-].[Cl-].[Ir+3]
1
–
5
of
5
results
1
Iridium(III) chloride, anhydrous, Premion™, 99.99% (metals basis), Ir 63.9% min
CAS: 10025-83-9 Molecular Formula: Cl3Ir Molecular Weight (g/mol): 298.567 MDL Number: MFCD00011063 InChI Key: DANYXEHCMQHDNX-UHFFFAOYSA-K PubChem CID: 25563 IUPAC Name: trichloroiridium SMILES: Cl[Ir](Cl)Cl
2
VeriSpec™ Iridium Standard for ICP 1000 ppm in 10% HCl, Ricca Chemical
CAS: 10025-83-9 Molecular Formula: Cl3Ir Molecular Weight (g/mol): 298.567 InChI Key: DANYXEHCMQHDNX-UHFFFAOYSA-K PubChem CID: 25563 IUPAC Name: trichloroiridium SMILES: Cl[Ir](Cl)Cl
4
VeriSpec™ Iridium Standard for ICP 100 ppm in 2% HCl, Ricca Chemical
CAS: 10025-83-9 Molecular Formula: Cl3Ir Molecular Weight (g/mol): 298.567 InChI Key: DANYXEHCMQHDNX-UHFFFAOYSA-K PubChem CID: 25563 IUPAC Name: trichloroiridium SMILES: Cl[Ir](Cl)Cl