missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Iridium Standard for ICP 1000 ppm in 10% HCl, Ricca Chemical
Description
- 1000 ppm in 10% HCl
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 88.04%, 9.80%, 0.16% |
| CAS Max % | 91.64%, 10.20%, 0.16% |
| Specific Gravity | 1 |
| Physical Form | Liquid |
| UN Number | UN1789 |
| Melting Point | Approximately 0°C |
| Type | Standard |
| Quantity | 250 mL |
| Grade | High Purity |