missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Indium Standard for ICP/MS 100 ppm in 2% HNO3, Ricca Chemical

Catalog No. RV010527100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010527100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010527100 Supplier Ricca Chemical Company Supplier No. RV010527100N
Only null left

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 13465-14-0 , 7697-37-2 , 7732-18-5
Molecular Formula H2InN3O10
Molecular Weight (g/mol) 318.845
InChI Key YZZFBYAKINKKFM-UHFFFAOYSA-N
PubChem CID 3084148
IUPAC Name indium(3+);trinitrate;hydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[In+3]

Specifications

Packaging Natural LDPE Bottle
CAS Min % 96.01%, 1.96%, 0.03%
CAS Max % 99.93%, 2.04%, 0.03%
Specific Gravity 1
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 100 mL
Grade High Purity
Chemical Name or Material Indium ICP-MS Standard
Show More Show Less