accessibility menu, dialog, popup

UserName

H2OInONOOONOOONOO

CAS RN 13465-14-0

IUPAC Name: indium(3+) hydrate trinitrate
Synonyms: indium(3+) hydrate trinitrate
Molecular Weight (g/mol): 318.85
Molecular Formula: H2InN3O10
InChi Key: YZZFBYAKINKKFM-UHFFFAOYSA-N
SMILES: O.[In+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results