missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Germanium Standard for ICP 1000 ppm in 5% HNO3/HF, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

$458.20 - $1175.67

Chemical Identifiers

CAS 1310-53-8
Molecular Formula GeO2
Molecular Weight (g/mol) 104.628
InChI Key YBMRDBCBODYGJE-UHFFFAOYSA-N
PubChem CID 14796
IUPAC Name dioxogermane
SMILES O=[Ge]=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RV010282100
View Documents
Ricca Chemical Company
RV010282100N
100 mL
Each for $458.20
Only null left
 
RV010282250
View Documents
Ricca Chemical Company
RV010282250N
250 mL
Each for $822.33
Only null left
 
RV010282500
View Documents
Ricca Chemical Company
RV010282500N
500 mL
Each for $1,175.67
Only null left
 
Description

Description

  • 1000 ppm in 5% HNO3/HF
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Chemical Identifiers

1310-53-8
104.628
14796
O=[Ge]=O
GeO2
YBMRDBCBODYGJE-UHFFFAOYSA-N
dioxogermane

Specifications

Natural LDPE Bottle
91.98%,4.90%,0.98%,0.14%
1
GeO2
Approximately 0°C
YBMRDBCBODYGJE-UHFFFAOYSA-N
dioxogermane
100 mL
High Purity
1310-53-8 , 7697-37-2 , 7664-39-3 , 7732-18-5
95.74%,5.10%,1.02%,0.14%
Liquid
UN2922
Standard
O=[Ge]=O
104.628
14796
Videos
SDS
Documents

Documents