CAS RN 1310-53-8
Germanium(IV) oxide, Puratronic™, 99.999% (metals basis)
CAS: 1310-53-8 Molecular Formula: GeO2 Molecular Weight (g/mol): 104.628 MDL Number: MFCD00011030 InChI Key: YBMRDBCBODYGJE-UHFFFAOYSA-N Synonym: germanium dioxide,germania,germanic acid,germanium oxide,germanium iv oxide,germanic oxide crystalline,dioxogermanium,germanic oxide,wln: ge o2 PubChem CID: 14796 IUPAC Name: dioxogermane SMILES: O=[Ge]=O
Germanium dioxide, 99.999%, (trace metal basis), electronic grade
CAS: 1310-53-8 Molecular Formula: GeO2 Molecular Weight (g/mol): 104.59 MDL Number: MFCD00011030 InChI Key: YBMRDBCBODYGJE-UHFFFAOYSA-N Synonym: germanium dioxide,germania,germanic acid,germanium oxide,germanium iv oxide,germanic oxide crystalline,dioxogermanium,germanic oxide,wln: ge o2 PubChem CID: 14796 IUPAC Name: dioxogermane SMILES: O=[Ge]=O
Germanium(IV) oxide, 99.99% (metals basis)
CAS: 1310-53-8 Molecular Formula: GeO2 Molecular Weight (g/mol): 104.628 MDL Number: MFCD00011030 InChI Key: YBMRDBCBODYGJE-UHFFFAOYSA-N Synonym: germanium dioxide,germania,germanic acid,germanium oxide,germanium iv oxide,germanic oxide crystalline,dioxogermanium,germanic oxide,wln: ge o2 PubChem CID: 14796 IUPAC Name: dioxogermane SMILES: O=[Ge]=O
VeriSpec™ Germanium Standard for ICP 1000 ppm in 5% HNO3/HF, Ricca Chemical
CAS: 1310-53-8 Molecular Formula: GeO2 Molecular Weight (g/mol): 104.628 InChI Key: YBMRDBCBODYGJE-UHFFFAOYSA-N PubChem CID: 14796 IUPAC Name: dioxogermane SMILES: O=[Ge]=O
VeriSpec™ Germanium Standard for ICP/MS 10 ppm in 2% HNO3/HF, Ricca Chemical
CAS: 1310-53-8 Molecular Formula: GeO2 Molecular Weight (g/mol): 104.628 InChI Key: YBMRDBCBODYGJE-UHFFFAOYSA-N PubChem CID: 14796 IUPAC Name: dioxogermane SMILES: O=[Ge]=O
VeriSpec™ Germanium (Ge) Standard for AAS 1000 ppm in 5% HNO3/1% HF, Ricca Chemical
CAS: 1310-53-8 Molecular Formula: GeO2 Molecular Weight (g/mol): 104.628 InChI Key: YBMRDBCBODYGJE-UHFFFAOYSA-N PubChem CID: 14796 IUPAC Name: dioxogermane SMILES: O=[Ge]=O
VeriSpec™ Germanium Standard for ICP/MS 100 ppm in 2% HNO3, Ricca Chemical
CAS: 1310-53-8 Molecular Formula: GeO2 Molecular Weight (g/mol): 104.628 InChI Key: YBMRDBCBODYGJE-UHFFFAOYSA-N PubChem CID: 14796 IUPAC Name: dioxogermane SMILES: O=[Ge]=O