missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Germanium Standard for ICP 1000 ppm in 5% HNO3/HF, Ricca Chemical

Catalog No. RV010282500
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010282500 500 mL
RV010282100 100 mL
RV010282250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RV010282500 Supplier Ricca Chemical Company Supplier No. RV010282500N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 5% HNO3/HF
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 1310-53-8 , 7697-37-2 , 7664-39-3 , 7732-18-5
Molecular Formula GeO2
Molecular Weight (g/mol) 104.628
InChI Key YBMRDBCBODYGJE-UHFFFAOYSA-N
PubChem CID 14796
IUPAC Name dioxogermane
SMILES O=[Ge]=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 91.98%, 4.90%, 0.98%, 0.14%
CAS Max % 95.74%, 5.10%, 1.02%, 0.14%
Specific Gravity 1
Physical Form Liquid
UN Number UN2922
Melting Point Approximately 0°C
Type Standard
Quantity 500 mL
Grade High Purity