missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Germanium (Ge) Standard for AAS 1000 ppm in 5% HNO3/1% HF, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

$316.50 - $681.20

Chemical Identifiers

CAS 1310-53-8
Molecular Formula GeO2
Molecular Weight (g/mol) 104.628
InChI Key YBMRDBCBODYGJE-UHFFFAOYSA-N
PubChem CID 14796
IUPAC Name dioxogermane
SMILES O=[Ge]=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RV010022100
View Documents
Ricca Chemical Company
RV010022100N
100 mL
Each for $316.50
Only null left
 
RV010022500
View Documents
Ricca Chemical Company
RV010022500N
500 mL
Each for $681.20
Only null left
 
Description

Description

  • 1000 ppm in 5% HNO3/1% HF
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Chemical Identifiers

1310-53-8
104.628
14796
O=[Ge]=O
GeO2
YBMRDBCBODYGJE-UHFFFAOYSA-N
dioxogermane

Specifications

Natural LDPE Bottle
91.98%,4.90%,0.98%,0.14%
1.01
GeO2
Approximately 0°C
O=[Ge]=O
104.628
14796
1310-53-8 , 7697-37-2 , 7664-39-3 , 7732-18-5
95.74%,5.10%,1.02%,0.14%
Liquid
UN2922
YBMRDBCBODYGJE-UHFFFAOYSA-N
dioxogermane
100 mL
High Purity
Videos
SDS
Documents

Documents