missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Germanium (Ge) Standard for AAS 1000 ppm in 5% HNO3/1% HF, Ricca Chemical
Description
- 1000 ppm in 5% HNO3/1% HF
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 91.98%, 4.90%, 0.98%, 0.14% |
| CAS Max % | 95.74%, 5.10%, 1.02%, 0.14% |
| Specific Gravity | 1.01 |
| Physical Form | Liquid |
| UN Number | UN2922 |
| Melting Point | Approximately 0°C |
| Type | Single-element Standard Solution |
| Quantity | 500 mL |
| Grade | High Purity |
| Show More |