missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Dysprosium (Dy) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

$315.69 - $642.44

Chemical Identifiers

CAS 10031-49-9
Molecular Formula DyH10N3O14
Molecular Weight (g/mol) 438.59
MDL Number MFCD00149683
InChI Key NOTQUFQJAWMLCE-UHFFFAOYSA-N
IUPAC Name dysprosium(3+) pentahydrate trinitrate
SMILES O.O.O.O.O.[Dy+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RV010016100
View Documents
Ricca Chemical Company
RV010016100N
100 mL
Each for $315.69
Only null left
 
RV010016500
View Documents
Ricca Chemical Company
RV010016500N
500 mL
Each for $642.44
Only null left
 
Description

Description

  • 1000 ppm in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Chemical Identifiers

10031-49-9
438.59
NOTQUFQJAWMLCE-UHFFFAOYSA-N
O.O.O.O.O.[Dy+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
DyH10N3O14
MFCD00149683
dysprosium(3+) pentahydrate trinitrate

Specifications

Natural LDPE Bottle
95.78%,1.96%,0.26%
1.01
DyH10N3O14
Approximately 0°C
O.O.O.O.O.[Dy+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
438.59
High Purity
10031-49-9,7697-37-2,7732-18-5
99.68%,2.04%,0.28%
Liquid
MFCD00149683
NOTQUFQJAWMLCE-UHFFFAOYSA-N
dysprosium(3+) pentahydrate trinitrate
100 mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'