missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Cesium Standard for ICP/MS 10 ppm in 2% HNO3, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010583100N

Catalog No. RV010583100


Only null left
Explore available promotions

Description

Description

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Cesium ICP-MS Standard
7789-18-6 , 7697-37-2 , 7732-18-5
99.96%,2.04%,0.00%
Liquid
Colorless
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Calibration Standard
NLSCHDZTHVNDCP-UHFFFAOYSA-N
cesium;nitrate
100 mL
High Purity
Natural LDPE Bottle
96.04%,1.96%,0.00%
1
CsNO3
Approximately 0°C
Miscible with water
Standard
[N+](=O)([O-])[O-].[Cs+]
194.909
62674

Chemical Identifiers

7789-18-6
194.909
62674
[N+](=O)([O-])[O-].[Cs+]
CsNO3
NLSCHDZTHVNDCP-UHFFFAOYSA-N
cesium;nitrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions