missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Cesium Standard for ICP 10 g/L in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010446100N
Description
- 10 g/L in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Cesium ICP Standard | |
| 7789-18-6 , 7697-37-2 , 7732-18-5 | |
| 98.46%,2.04%,1.50% | |
| Liquid | |
| Colorless | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Calibration Standard | |
| NLSCHDZTHVNDCP-UHFFFAOYSA-N | |
| cesium;nitrate | |
| 100 mL | |
| High Purity |
| Natural LDPE Bottle | |
| 94.60%,1.96%,1.44% | |
| 1 | |
| CsNO3 | |
| Approximately 0°C | |
| Miscible with water | |
| Standard | |
| [N+](=O)([O-])[O-].[Cs+] | |
| 194.909 | |
| 62674 |