missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Calcium Standard for ICP 10 g/L in 5% HNO3, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010371500N

 View more versions of this product

Catalog No. RV010371500


Only null left

Description

Description

  • 10 g/L in 5% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Natural Poly Bottle
89.09%,4.90%,4.01%
1
CaN2O6
Approximately 0°C
ZCCIPPOKBCJFDN-UHFFFAOYSA-N
calcium;dinitrate
500 mL
CHEBI:64205
10124-37-5 , 7697-37-2 , 7732-18-5
92.73%,5.10%,4.17%
Liquid
UN3264
Standard
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ca+2]
164.086
24963
High Purity

Chemical Identifiers

10124-37-5
164.086
24963
calcium;dinitrate
CaN2O6
ZCCIPPOKBCJFDN-UHFFFAOYSA-N
CHEBI:64205
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ca+2]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions