missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Calcium Standard for ICP 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$461.10 - $1069.67
Chemical Identifiers
| CAS | 10124-37-5 |
|---|---|
| Molecular Formula | CaN2O6 |
| Molecular Weight (g/mol) | 164.086 |
| InChI Key | ZCCIPPOKBCJFDN-UHFFFAOYSA-N |
| PubChem CID | 24963 |
| ChEBI | CHEBI:64205 |
| IUPAC Name | calcium;dinitrate |
| SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ca+2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010335100
|
Ricca Chemical Company
RV010335100N |
100 mL |
Each for $461.10
|
|
|||||
|
RV010335250
|
Ricca Chemical Company
RV010335250N |
250 mL |
Each for $751.53
|
|
|||||
|
RV010335500
|
Ricca Chemical Company
RV010335500N |
500 mL |
Each for $1,069.67
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 10124-37-5 | |
| 164.086 | |
| 24963 | |
| calcium;dinitrate |
| CaN2O6 | |
| ZCCIPPOKBCJFDN-UHFFFAOYSA-N | |
| CHEBI:64205 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ca+2] |
Specifications
| Natural LDPE Bottle | |
| 95.64%,1.96%,0.40% | |
| 1 | |
| CaN2O6 | |
| Standard | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Ca+2] | |
| 164.086 | |
| 24963 | |
| High Purity |
| 10124-37-5 , 7697-37-2 , 7732-18-5 | |
| 99.54%,2.04%,0.42% | |
| Liquid | |
| Approximately 0°C | |
| ZCCIPPOKBCJFDN-UHFFFAOYSA-N | |
| calcium;dinitrate | |
| 100 mL | |
| CHEBI:64205 |