missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Cadmium (Cd) Standard for AAS 1000 ppm in 2% HCl, Ricca Chemical
Description
- 1000 ppm in 2% HCl
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 95.88%, 1.96%, 0.16% |
| CAS Max % | 99.80%, 2.04%, 0.16% |
| Specific Gravity | 1.01 |
| Physical Form | Liquid |
| Melting Point | Approximately 0°C |
| Type | Single-element Standard Solution |
| Quantity | 500 mL |
| Grade | High Purity |
| Chemical Name or Material | Cadmium |