accessibility menu, dialog, popup

UserName

ClClCd

CAS RN 10108-64-2

IUPAC Name: cadmium(2+) dichloride
Synonyms: cadmium chloride
Molecular Weight (g/mol): 183.31
Molecular Formula: CdCl2
InChi Key: YKYOUMDCQGMQQO-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Cd++]
Grade 
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
Percent Purity 
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (11)
  • (1)
Physical Form 
  • (2)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (4)
  • (9)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (11)
  • (1)
  • (2)
  • (7)