missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Bismuth Standard for ICP 10 g/L in 10% HNO3, Ricca Chemical

Catalog No. RV010370100
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010370100 100 mL
RV010370500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010370100 Supplier Ricca Chemical Company Supplier No. RV010370100N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 10 g/L in 10% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 10035-06-0 , 7697-37-2 , 7732-18-5
Molecular Formula BiH10N3O14
Molecular Weight (g/mol) 485.07
MDL Number MFCD00149157
InChI Key FBXVOTBTGXARNA-UHFFFAOYSA-N
PubChem CID 160911
IUPAC Name bismuth(3+) pentahydrate trinitrate
SMILES O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 86.22%, 9.80%, 1.98%
CAS Max % 89.74%, 10.20%, 2.06%
Specific Gravity 1
Physical Form Liquid
UN Number UN3264
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Grade High Purity