missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Beryllium Standard for ICP 1000 ppm in 2% HNO3/HF, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$988.37 - $2306.47
Chemical Identifiers
| CAS | 13597-99-4 |
|---|---|
| Molecular Formula | BeN2O6 |
| Molecular Weight (g/mol) | 133.02 |
| InChI Key | RFVVBBUVWAIIBT-UHFFFAOYSA-N |
| PubChem CID | 26126 |
| IUPAC Name | beryllium;dinitrate |
| SMILES | [Be+2].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010334100
|
Ricca Chemical Company
RV010334100N |
100 mL |
Each for $988.37
|
|
|||||
|
RV010334250
|
Ricca Chemical Company
RV010334250N |
250 mL |
Each for $1,599.97
|
|
|||||
|
RV010334500
|
Ricca Chemical Company
RV010334500N |
500 mL |
Each for $2,306.47
|
|
|||||
Description
- 1000 ppm in 2% HNO3/HF
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 13597-99-4 | |
| 133.02 | |
| 26126 | |
| [Be+2].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-] |
| BeN2O6 | |
| RFVVBBUVWAIIBT-UHFFFAOYSA-N | |
| beryllium;dinitrate |
Specifications
| Natural LDPE Bottle | |
| 94.10%,1.96%,1.45%,0.49% | |
| 1 | |
| BeN2O6 | |
| Standard | |
| [Be+2].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-] | |
| 133.02 | |
| 26126 |
| 13597-99-4 , 7697-37-2 , 7664-39-3 , 7732-18-5 | |
| 97.94%,2.04%,1.51%,0.51% | |
| Liquid | |
| Approximately 0°C | |
| RFVVBBUVWAIIBT-UHFFFAOYSA-N | |
| beryllium;dinitrate | |
| 100 mL | |
| High Purity |