CAS RN 13597-99-4
CAS RN 13597-99-4
IUPAC Name:
beryllium(2+) dinitrate
Synonyms:
beryllium nitrate
Molecular Weight (g/mol):
133.02
Molecular Formula:
BeN2O6
InChi Key:
RFVVBBUVWAIIBT-UHFFFAOYSA-N
SMILES:
[Be++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
1
–
4
of
4
results
3
VeriSpec™ Beryllium (Be) Standard for AAS 1000 ppm in 2% HNO3/1% HF, Ricca Chemical
CAS: 13597-99-4 Molecular Formula: BeN2O6 Molecular Weight (g/mol): 133.02 InChI Key: RFVVBBUVWAIIBT-UHFFFAOYSA-N PubChem CID: 26126 IUPAC Name: beryllium;dinitrate SMILES: [Be+2].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-]
4
VeriSpec™ Beryllium Standard for ICP 1000 ppm in 2% HNO3/HF, Ricca Chemical
CAS: 13597-99-4 Molecular Formula: BeN2O6 Molecular Weight (g/mol): 133.02 InChI Key: RFVVBBUVWAIIBT-UHFFFAOYSA-N PubChem CID: 26126 IUPAC Name: beryllium;dinitrate SMILES: [Be+2].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-]