missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Barium (Ba) Standard for AAS 1000 ppm in 2% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$249.47 - $439.50
Chemical Identifiers
| CAS | 10022-31-8 |
|---|---|
| Molecular Formula | BaN2O6 |
| Molecular Weight (g/mol) | 261.34 |
| MDL Number | MFCD00003442 |
| InChI Key | IWOUKMZUPDVPGQ-UHFFFAOYSA-N |
| PubChem CID | 24798 |
| IUPAC Name | barium(2+) dinitrate |
| SMILES | [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010066100
|
Ricca Chemical Company
RV010066100N |
100 mL |
Each for $249.47
|
|
|||||
|
RV010066500
|
Ricca Chemical Company
RV010066500N |
500 mL |
Each for $439.50
|
|
|||||
Description
- 1000 ppm in 2% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 10022-31-8 | |
| 261.34 | |
| IWOUKMZUPDVPGQ-UHFFFAOYSA-N | |
| barium(2+) dinitrate |
| BaN2O6 | |
| MFCD00003442 | |
| 24798 | |
| [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
| Natural LDPE Bottle | |
| 95.85%,1.96%,0.19% | |
| 1.01 | |
| BaN2O6 | |
| Approximately 0°C | |
| [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 261.34 | |
| 24798 |
| 10022-31-8 , 7697-37-2 , 7732-18-5 | |
| 99.77%,2.04%,0.19% | |
| Liquid | |
| MFCD00003442 | |
| IWOUKMZUPDVPGQ-UHFFFAOYSA-N | |
| barium(2+) dinitrate | |
| 100 mL | |
| High Purity |