missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Antimony Standard for ICP 1000 ppm in 5% HNO3/HF, Ricca Chemical
Description
- 1000 ppm in 5% HNO3/HF
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 91.92%, 4.90%, 0.98%, 0.20% |
| CAS Max % | 95.68%, 5.10%, 1.02%, 0.20% |
| Specific Gravity | 1 |
| Physical Form | Liquid |
| UN Number | UN2922 |
| Melting Point | Approximately 0°C |
| Type | Standard |
| Quantity | 500 mL |
| Grade | High Purity |