missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Aluminum Standard for ICP 1000 ppm in 2% HCl, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010261500N
Description
- 1000 ppm in 2% HCl
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Natural Poly Bottle | |
| 95.56%,1.96%,0.48% | |
| 1 | |
| AlCl3 | |
| Approximately 0°C | |
| VSCWAEJMTAWNJL-UHFFFAOYSA-K | |
| aluminum(3+) trichloride | |
| 500 mL |
| 7446-70-0 , 7647-01-0 , 7732-18-5 | |
| 99.46%,2.04%,0.50% | |
| Liquid | |
| MFCD00003422 | |
| Standard | |
| [Al+3].[Cl-].[Cl-].[Cl-] | |
| 133.33 | |
| High Purity |
Chemical Identifiers
| 7446-70-0 | |
| 133.33 | |
| VSCWAEJMTAWNJL-UHFFFAOYSA-K | |
| [Al+3].[Cl-].[Cl-].[Cl-] |
| AlCl3 | |
| MFCD00003422 | |
| aluminum(3+) trichloride |