missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Aluminum Standard for ICP 1000 ppm in 2% HCl, Ricca Chemical

Catalog No. p-7073821
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010261100 100 mL
RV010261250 250 mL
RV010261500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RV010261100 Supplier Ricca Chemical Company Supplier No. RV010261100N
Only null left

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 2% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7446-70-0 , 7647-01-0 , 7732-18-5
Molecular Formula AlCl3
Molecular Weight (g/mol) 133.33
MDL Number MFCD00003422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
IUPAC Name aluminum(3+) trichloride
SMILES [Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Packaging Natural LDPE Bottle
CAS Min % 95.56%, 1.96%, 0.48%
CAS Max % 99.46%, 2.04%, 0.50%
Specific Gravity 1
Physical Form Liquid
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Grade High Purity