missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Aluminum Standard for ICP 10 g/L in 5% HNO3, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Supplier: Ricca Chemical Company RV010434250N
Description
- 10 g/L in 5% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
| Natural Poly Bottle | |
| 79.48%,13.62%,4.90% | |
| 1 | |
| UN3264 | |
| Standard | |
| High Purity |
| 13473-90-0 , 7697-37-2 , 7732-18-5 | |
| 82.72%,14.18%,5.10% | |
| Liquid | |
| Approximately 0°C | |
| 250 mL |
Chemical Identifiers
| 13473-90-0 |