missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Aluminum Standard for ICP 10 g/L in 5% HNO3, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

$847.17 - $3756.30

Chemical Identifiers

CAS 13473-90-0
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RV010434100
View Documents
Ricca Chemical Company
RV010434100N
100 mL
Each for $847.17
Only null left
 
RV010434250
View Documents
Ricca Chemical Company
RV010434250N
250 mL
Each for $2,201.53
Only null left
 
RV010434500
View Documents
Ricca Chemical Company
RV010434500N
500 mL
Each for $3,756.30
Only null left
 
Description

Description

  • 10 g/L in 5% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Chemical Identifiers

13473-90-0

Specifications

Natural LDPE Bottle
79.48%,13.62%,4.90%
1
UN3264
Standard
High Purity
13473-90-0 , 7697-37-2 , 7732-18-5
82.72%,14.18%,5.10%
Liquid
Approximately 0°C
100 mL
Videos
SDS
Documents

Documents