missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Aluminum Standard for ICP 10 g/L in 5% HCl, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Supplier:  Ricca Chemical Company RV010364500N

 View more versions of this product

Catalog No. RV010364500


Only null left

Description

Description

  • 10 g/L in 5% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Natural Poly Bottle
88.26%,4.90%,4.84%
1
AlCl3
UN3264
Standard
[Al+3].[Cl-].[Cl-].[Cl-]
133.33
High Purity
7446-70-0 , 7647-01-0 , 7732-18-5
91.86%,5.10%,5.04%
Liquid
MFCD00003422
Approximately 0°C
VSCWAEJMTAWNJL-UHFFFAOYSA-K
aluminum(3+) trichloride
500 mL

Chemical Identifiers

7446-70-0
133.33
VSCWAEJMTAWNJL-UHFFFAOYSA-K
[Al+3].[Cl-].[Cl-].[Cl-]
AlCl3
MFCD00003422
aluminum(3+) trichloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions