missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ Aluminum Standard for ICP 10 g/L in 5% HCl, Ricca Chemical
Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
$847.17 - $3756.30
Chemical Identifiers
| CAS | 7446-70-0 |
|---|---|
| Molecular Formula | AlCl3 |
| Molecular Weight (g/mol) | 133.33 |
| MDL Number | MFCD00003422 |
| InChI Key | VSCWAEJMTAWNJL-UHFFFAOYSA-K |
| IUPAC Name | aluminum(3+) trichloride |
| SMILES | [Al+3].[Cl-].[Cl-].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RV010364100
|
Ricca Chemical Company
RV010364100N |
100 mL |
Each for $847.17
|
|
|||||
|
RV010364500
|
Ricca Chemical Company
RV010364500N |
500 mL |
Each for $3,756.30
|
|
|||||
Description
- 10 g/L in 5% HCl
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Chemical Identifiers
| 7446-70-0 | |
| 133.33 | |
| VSCWAEJMTAWNJL-UHFFFAOYSA-K | |
| [Al+3].[Cl-].[Cl-].[Cl-] |
| AlCl3 | |
| MFCD00003422 | |
| aluminum(3+) trichloride |
Specifications
| Natural LDPE Bottle | |
| 88.26%,4.90%,4.84% | |
| 1 | |
| AlCl3 | |
| UN3264 | |
| Standard | |
| [Al+3].[Cl-].[Cl-].[Cl-] | |
| 133.33 | |
| High Purity |