Learn More
Staco Energy™ Variable Transformers
Compact, portable, cord-and-plug type unit
Supplier: Staco Energy™ 3PN1010B
Description
Compact, portable, cord-and-plug type unit
- Three-prong line cord and receptacle
- Illuminated ON/OFF switch
- Housed in a rugged, ventilated steel enclosure
- 50/60Hz
- Select output voltage from 0-117% line voltage
- Increase output voltage with a clockwise rotation
Identifiants chimiques
| Variable Transformer | |
| ±5% | |
| Three prong plug |
| Steel | |
| 50/60 Hz |
Spécifications
| Variable Transformer | |
| 6.96 in. | |
| Steel | |
| ±5% | |
| 120 V, 50/60 Hz | |
| 13.9 cm | |
| Three prong plug | |
| 4.64 kg |
| Steel | |
| 17.68 cm | |
| 120 V | |
| 10 A | |
| 5.5 in. | |
| 50/60 Hz | |
| 10.25 lb. | |
| 11.7 cm |