missing translation for 'onlineSavingsMsg'
Learn More

Vanadyl Sulfate, Hydrate, 97%, Spectrum™ Chemical

Numéro de catalogue. 18614649
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
20 kg
Packaging:
Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
18614649 25 g Poly Bottle
18614647 100 g Poly Bottle
18614651 500 g Poly Bottle
18614650 5 kg Poly Pail
18614648 20 kg Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Numéro de catalogue. 18614649 Fournisseur Spectrum Chemical Mfg Cor Code fournisseur V102025GM
Il en reste null

Chemical Identifiers

CAS 123334-20-3

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 97%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.