missing translation for 'onlineSavingsMsg'
Learn More

Vanadium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLV22X

 View more versions of this product

Catalog No. PLV2-2X


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
V1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Vanadium Standard
7803-55-6 , 7697-37-2 , 7732-18-5
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
1.0,15.0,84.0
MFCD00011430
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
YBVKNHXQSRDWAA-UHFFFAOYSA-M
2% HNO3
116.98
516859
Matrix    Concentration    Cat. No.
2% HNO3    1000μg/mL    PLV2-2Y, -2X
15% HNO3    10,000    PLV4-3Y, -3X
2% HCl    1000μg/mL    PLV1-2X
15% HCl    10,000    PLV3-3X
Triple-leached LDPE bottle
Assurance Certificate of Analysis reports up 30 elements at ppb levels.
1.0,15.0,84.0
H4NO3V
AA, ICP
Certified Reference Material
N.O[V](=O)=O
vanadiumoylol amine
500 mL
Plasma

Chemical Identifiers

7803-55-6
116.98
YBVKNHXQSRDWAA-UHFFFAOYSA-M
N.O[V](=O)=O
H4NO3V
MFCD00011430
516859
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation. May cause respiratory irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only