missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanadium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$305.97 - $611.94
Chemical Identifiers
| CAS | 7803-55-6 |
|---|---|
| Molecular Formula | H4NO3V |
| Molecular Weight (g/mol) | 116.98 |
| MDL Number | MFCD00011430 |
| InChI Key | YBVKNHXQSRDWAA-UHFFFAOYSA-M |
| PubChem CID | 516859 |
| SMILES | N.O[V](=O)=O |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PLV2-2Y
|
SPEX CertiPrep Group LLC
PLV22Y |
125 mL | Triple-leached LDPE bottle |
Each for $305.97
|
|
||||
|
PLV2-2X
|
SPEX CertiPrep Group LLC
PLV22X |
500 mL | Triple-leached LDPE bottle |
Each for $611.94
|
|
||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| V | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 7803-55-6 | |
| 116.98 | |
| YBVKNHXQSRDWAA-UHFFFAOYSA-M | |
| N.O[V](=O)=O |
| H4NO3V | |
| MFCD00011430 | |
| 516859 |
Specifications
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up 30 elements at ppb levels. | |
| 1.0,15.0,84.0 | |
| H4NO3V | |
| AA, ICP | |
| Certified Reference Material | |
| N.O[V](=O)=O | |
| azanium;oxido(dioxo)vanadium | |
| 125 mL | |
| Plasma | |
| Vanadium Standard |
| 7803-55-6 , 7697-37-2 , 7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 1.0,15.0,84.0 | |
| MFCD00011430 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| YBVKNHXQSRDWAA-UHFFFAOYSA-M | |
| 2% HNO3 | |
| 116.98 | |
| 516859 | |
| Matrix Concentration Cat. No. 2% HNO3 1000μg/mL PLV2-2Y, -2X 15% HNO3 10,000 PLV4-3Y, -3X 2% HCl 1000μg/mL PLV1-2X 15% HCl 10,000 PLV3-3X |
Safety and Handling
Causes skin irritation, serious eye irritation. May cause respiratory irritation
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only