Learn More
Vanadium AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg V), LabChem™
$87.07 - $182.57
Chemical Identifiers
| CAS | 7803-55-6 |
|---|---|
| Molecular Formula | H4NO3V |
| Molecular Weight (g/mol) | 116.98 |
| MDL Number | MFCD00011430 |
| InChI Key | YBVKNHXQSRDWAA-UHFFFAOYSA-M |
| PubChem CID | 516859 |
| IUPAC Name | vanadiumoylol amine |
| SMILES | N.O[V](=O)=O |
Chemical Identifiers
| 7803-55-6 | |
| 116.98 | |
| YBVKNHXQSRDWAA-UHFFFAOYSA-M | |
| vanadiumoylol amine |
| H4NO3V | |
| MFCD00011430 | |
| 516859 | |
| N.O[V](=O)=O |
Specifications
| Poly Bottle | |
| 95.76,4,0.24 | |
| Liquid | |
| 1000 ppm ±10 ppm (1mL = 1mg V) | |
| MFCD00011430 | |
| Green/Yellow | |
| Soluble in water | |
| N.O[V](=O)=O | |
| 116.98 | |
| 516859 | |
| Certified |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature