Learn More
Vanadium AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg V), LabChem™
Supplier: LabChem LC266401
Specifications
| Vanadium AA Standard | |
| Poly Bottle | |
| 95.76,4,0.24 | |
| Liquid | |
| 1000 ppm ±10 ppm (1mL = 1mg V) | |
| MFCD00011430 | |
| Green/Yellow | |
| Soluble in water | |
| N.O[V](=O)=O | |
| 116.98 | |
| 516859 | |
| Certified |
| 1000ppm (1mL = 1mg V) | |
| 7803-55-6 , 7697-37-2 , 7732-18-5 | |
| Passes Test | |
| H4NO3V | |
| NH4VO3 | |
| UN3264 | |
| 105°C | |
| YBVKNHXQSRDWAA-UHFFFAOYSA-M | |
| vanadiumoylol amine | |
| 500 mL | |
| 116.97 |
Chemical Identifiers
| 7803-55-6 | |
| 116.98 | |
| YBVKNHXQSRDWAA-UHFFFAOYSA-M | |
| vanadiumoylol amine |
| H4NO3V | |
| MFCD00011430 | |
| 516859 | |
| N.O[V](=O)=O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature