missing translation for 'onlineSavingsMsg'
Learn More

VAI Easy2Gown™ Mitcool 1700 Cleanroom Hoods

Catalog No. 19546428
Change view
Click to view available options
Size:
Large/X-Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19546428 Small/Medium
19547262 Large/X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19546428 Supplier VAI Supplier No. 1700PHUCS17017

VAI Easy2Gown™ Mitcool 1700 Cleanroom Hoods are single-use PPE with excellent chemical resistance

  • Folded in an inside-out fashion that essentially elimates the individual from contaminating the outside of the garment
  • Packaged to prevent operator contamination of the outside of the garment
  • Bonded polypropylene with laminate
  • Piped seams with reinforced stitching
  • Excellent chemical resistance

Specifications

Product Type Cleanroom Hood
Size Small/Medium
Material Polypropylene Laminate
Color White
Disposable Single-use
Seam Type Piped, Stitched
Sterility Sterile
Quantity 100/Cs.
Weight (English) 2.5 oz./sq. yd
Includes Shipped with Lot Specific Documentation
For Use With (Application) Personal Protective Equipment (PPE)
Resistance Chemicals
Hood Type Eye Openings
Cleanroom Class Class 100-1000 (ISO 5-6)
Show More Show Less