Learn More
Greiner Bio-One VACUETTE™ Urine CCM Tubes
VACUETTE™ Urine CCM Tubes offer immediate sample stabilization for improved preanalytics
Supplier:  Greiner Bio-One 454486
 Afficher plus de versions de ce produit
Numéro de catalogue. 22040550
Il en reste null
Description
Description
  • Count control and minnitol powder: Additive stabilizes the bacterial count and parameters for up to 48 hours at room temperature.
    • Stabilizer contains boric acid, sodium tetraborate, socium formate and mannitol
    • Urine specimen does not require refrigeration.
  • Suitable to extended processing and transport times, and for microbiological diagnosis
  • Leak-proof
  • PET plastic is virutally unbreakable
  • Feature a vacuum for an optimized sample-to-stabilizer ratio
Spécifications
Identifiants chimiques
Urine Tube
Sterile
1200/Cs.
Yellow with Black Ring
Non-ridged
50 Pk.
Spécifications
Urine Tube
Sterile
Non-ridged
50 Pk.
Yellow with Black Ring
Round
1200/Cs.
4.0 mL
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS