Learn More
MSA™ V-Gard™ Visor
Numéro de catalogue. 19030078
Change view
Click to view available options
Lens Tint:
IR Shade 10
IR Shade 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Lens Tint
19030078 IR Shade 10
19030079 IR Shade 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 19030078 Fournisseur MSA™ Code fournisseur 10246714
Il en reste null
V-Gard polycarbonate shaded visors are thick and molded to protect against damage from impact, elevated temperatures, weld spatter and infrared (IR) radiation. Ideal for autogenous welding, metal pouring, gas soldering and light cutting/brazing, they’re engineered for heavy-duty use.
  • Thickness helps prevent pitting and warping from heat or weld spatter
  • Suitable for applications where welding, cutting and brazing hazards exist
  • V-Gard polycarbonate shaded visors are thick and molded to protect against damage from impact, elevated temperatures, weld spatter and infrared (IR) radiation
  • Ideal for autogenous welding, metal pouring, gas soldering and light cutting/brazing, they are engineered for heavy-duty use
Spécifications
Lens Coating Anti-scratch, Anti-fog, Impact-resistant
Lens Tint IR Shade 10
Product Type Visor
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection None