missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Uridine, 99%

CAS: 58-96-8 | C9H12N2O6 | 244.2 g/mol

Chemical Identifiers

CAS 58-96-8
Molecular Formula C9H12N2O6
Molecular Weight (g/mol) 244.2
MDL Number MFCD00006526
InChI Key DRTQHJPVMGBUCF-DHRBYNEYSA-N
Synonym uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urd
PubChem CID 89495556
IUPAC Name 1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC140770010
View Documents Promotion Details
Thermo Scientific Chemicals
140770010
1 g Glass bottle N/A
Only null left
 
AC140770250
View Documents Promotion Details
Thermo Scientific Chemicals
140770250
25 g Glass bottle N/A
Only null left
 
AC140771000
View Documents Promotion Details
Thermo Scientific Chemicals
140771000
100 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

58-96-8
244.2
DRTQHJPVMGBUCF-DHRBYNEYSA-N
89495556
C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O
C9H12N2O6
MFCD00006526
uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urd
1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione

Specifications

58-96-8
162.0°C to 170.0°C
1% max. (105°C, 2 hrs)
98.5% min. (HPLC)
MFCD00006526
15, 10062
uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urd
DRTQHJPVMGBUCF-DHRBYNEYSA-N
95% min. (c=1, H2O, 430nm) 1cm cell
244.2
244.2
Crystalline Powder
0.72 to 0.76 (A250/A260), 0.34 to 0.38 (A280/A260)
White
Authentic
C9H12N2O6
31, 23
+ 8.40
98% min. (lambda max. 262nm,pH 7) molar absorptivity 10100+/-2%,on dry basis
C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O
1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
89495556
99%
Uridine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-407-5

SDS
Documents

Documents

RUO – Research Use Only