missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Uridine, 99%

Catalog No. AC140770250
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC140770250 25 g Glass bottle
AC140770010 1 g Glass bottle
AC140771000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC140770250 Supplier Thermo Scientific Chemicals Supplier No. 140770250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58-96-8 | C9H12N2O6 | 244.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58-96-8
Molecular Formula C9H12N2O6
Molecular Weight (g/mol) 244.2
MDL Number MFCD00006526
InChI Key DRTQHJPVMGBUCF-DHRBYNEYSA-N
Synonym uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urd
PubChem CID 89495556
IUPAC Name 1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O

Specifications

Absorbance 0.72 to 0.76 (A250/A260), 0.34 to 0.38 (A280/A260)
Melting Point 162.0°C to 170.0°C
Color White
Loss on Drying 1% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 31, 23
Merck Index 15, 10062
Specific Rotation + 8.40
Spectrophotometry 98% min. (lambda max. 262nm,pH 7) molar absorptivity 10100+/-2%,on dry basis
Transmittance 95% min. (c=1, H2O, 430nm) 1cm cell
Formula Weight 244.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Uridine
Show More Show Less
EINECSNumber 200-407-5

RUO – Research Use Only