missing translation for 'onlineSavingsMsg'
Learn More

Uranium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLU23X

 View more versions of this product

Catalog No. PLU2-3X


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
U10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Uranium Standard (Depleted)
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
[U]
uranium
500 mL
CHEBI:49936
Triple-leached LDPE bottle
7440-61-1 , 7697-37-2 , 7732-18-5
U
JFALSRSLKYAFGM-UHFFFAOYSA-N
5% HNO3
238.029
23989
Assurance

Chemical Identifiers

7440-61-1
238.029
23989
uranium
U
JFALSRSLKYAFGM-UHFFFAOYSA-N
CHEBI:49936
[U]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only