missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uranium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$622.56 - $1370.00
Chemical Identifiers
| CAS | 7440-61-1 |
|---|---|
| Molecular Formula | U |
| Molecular Weight (g/mol) | 238.029 |
| InChI Key | JFALSRSLKYAFGM-UHFFFAOYSA-N |
| PubChem CID | 23989 |
| ChEBI | CHEBI:49936 |
| IUPAC Name | uranium |
| SMILES | [U] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| U | 10,000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Triple-leached LDPE bottle | |
| 7440-61-1 , 7697-37-2 , 7732-18-5 | |
| U | |
| JFALSRSLKYAFGM-UHFFFAOYSA-N | |
| 5% HNO3 | |
| 238.029 | |
| 23989 | |
| Assurance |
| Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| [U] | |
| uranium | |
| 125 mL | |
| CHEBI:49936 | |
| Uranium Standard (Depleted) |
Safety and Handling
Causes skin irritation, serious eye irritation
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only