missing translation for 'onlineSavingsMsg'
Learn More

Universal Indicator, Bogens in Alcohol, Certified, LabChem™

$69.68 - $292.50

Specifications

Chemical Name or Material Universal Indicator
Name Note Bogens in Alcohol
CAS Max % 70,29.74,0.08,0.07,0.05
CAS 7732-18-5 , 76-59-5 , 547-58-0
Linear Formula C27H28Br2O5S
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
LC265007
View Documents
LabChem
LC265007
125 mL White Poly
Each for $69.68
Only null left
 
LC265001
View Documents
LabChem
LC265001
500 mL Amber Poly
Each for $195.75
Only null left
 
LC265002
View Documents
LabChem
LC265002
1 L Amber Poly
Each for $292.50
Only null left
 
Description

Description

Specifications

Specifications

Universal Indicator
70,29.74,0.08,0.07,0.05
C27H28Br2O5S
Soluble in water
624.38
Carbon dioxide; Carbon monoxide; Sulfur compounds; Hydrogen bromide
Liquid
Bogens in Alcohol
7732-18-5 , 76-59-5 , 547-58-0
UN1993
Passes Test
Certified
Green
Videos
SDS
Documents

Documents